missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Wood, Powder, Spectrum™ Chemical

Catalog No. 18609678
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609678 50 kg Fiber Drum
18609679 500 g Fiber Drum
18609677 2.5 kg Fiber Drum
18609676 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609678 Supplier Spectrum Chemical Mfg Cor Supplier No. C121750KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-44-0

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 50 kg
Molecular Weight (g/mol) 12.01
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Charcoal, Wood, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.