missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Coconut, Activated, 8-30 Mesh, Spectrum™ Chemical

Catalog No. p-7165313
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609674 500 g Amber Glass Bottle
18609675 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18609674 Supplier Spectrum Chemical Mfg Cor Supplier No. C1221500GM
Only null left

Chemical Identifiers

CAS 7440-44-0

Specifications

CAS Max % 1
Mesh Size 8 - 30
Packaging Amber Glass Bottle
Quantity 500 g
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.