missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Coconut, Activated, 8-30 Mesh, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Mesh Size | 8 - 30 |
| Packaging | Amber Glass Bottle |
| Quantity | 2.5 kg |
| Molecular Weight (g/mol) | 12.01 |
| Grade | Ungraded |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Charcoal, Coconut, Activated, 8-30 Mesh |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.