missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Activated, Powder, Spectrum™ Chemical

Catalog No. 18609671
Change view
Click to view available options
Quantity:
500 g
40 lb.
45 kg
Packaging:
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609671 45 kg Fiber Drum
18609672 500 g Fiber Drum
18609670 40 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609671 Supplier Spectrum Chemical Mfg Cor Supplier No. CH10345KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-44-0

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 45 kg
Molecular Weight (g/mol) 12.01
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Charcoal, Activated, Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.