missing translation for 'onlineSavingsMsg'
Learn More

CHAPS (White Crystalline Powder), Fisher BioReagents™

Catalog No. BP5711
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP5711 1 g Amber Glass
BP5715 5 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP5711 Supplier Fisher BioReagents Supplier No. FLBP571 1
CHAPS is a Zwitterionic detergent used for solubilizing membrane proteins.

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl) dimethylammonio]-1-propanesulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Absorbance 0.1 max. (1%) at 260nm, 0.1 max. (1%) at 280nm
Color White
Physical Form Solid
Melting Point 267°C
Quantity 1 g
Identification Pass Test
Packaging Amber Glass
Assay Percent Range ≥98 %
Merck Index 15, 2043
Formula Weight 614.9
Chemical Name or Material CHAPS
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage RT