missing translation for 'onlineSavingsMsg'
Learn More

CHAPS, 98+%

Catalog No. AAB2192703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2192703 1 g
AAB2192706 5 g
AAB2192714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2192703 Supplier Thermo Scientific Chemicals Supplier No. B2192703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHAPS, >-98%, C32H58N2O7S, CAS Number-75621-03-3, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 25g, 614.88, H315-H319-H335, GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. | CAS: 75621-03-3 | C32H58N2O7S | 614.883 g/mol

Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

Solubility
Slightly soluble in water.

Notes
Store in a cool. Incompatible with oxidizing agent, strong acid and bases.

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
MDL Number MFCD00012116
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Melting Point 156°C to 158°C (decomposition)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 614.88
Percent Purity ≥98%
Chemical Name or Material CHAPS
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only