Learn More
Kinematica Chamber and Piping Cleaner
Catalog No. KINCLEAN
Change view
Click to view available options
For Use With (Equipment):
Autoclave chamber or piping
Catalog No. For Use With (Equipment)
KINCLEAN Autoclave chamber or piping
1 options
Catalog No. KINCLEAN Supplier Kinematica Supplier No. KINCLEAN
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Maintains autoclave’s pristine condition with high-performance cleaner
  • Chamber Cleaner, recommends after 15 cycles
  • Maintain your autoclave's pristine condition with this high-performance cleaner
  • Ensuring consistent and effective sterilization cycles
Specifications
Autoclavable Autoclavable
Certifications/Compliance Quality Management System
For Use With (Application) Sterilization of any Chamber or piping for an Autoclave
For Use With (Equipment) Autoclave chamber or piping
Model KinClean
Weight (English) 12 oz.
Weight (Metric) 0.34 kg
Includes Box of 10 pouches