missing translation for 'onlineSavingsMsg'
Learn More

Chalcone 98.0+%, TCI America™

Catalog No. p-7124203
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C007125G 25 g
C0071100G 100 g
C0071500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C007125G Supplier TCI America Supplier No. C007125G
Only null left

Chemical Identifiers

CAS 94-41-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003082
InChI Key DQFBYFPFKXHELB-VAWYXSNFSA-N
Synonym chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone
PubChem CID 637760
ChEBI CHEBI:48965
IUPAC Name (2E)-1,3-diphenylprop-2-en-1-one
SMILES O=C(\C=C\C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color Yellow
Boiling Point 348°C
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chalcone
EINECSNumber (4)-0073
RTECSNumber FL6900000
TSCA Yes