missing translation for 'onlineSavingsMsg'
Learn More

Chalcone, 97%

Catalog No. p-84231
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159431000 100 g Plastic bottle
AC159435000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159431000 Supplier Thermo Scientific Chemicals Supplier No. 159431000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 94-41-7 | C15H12O | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-41-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003082
InChI Key DQFBYFPFKXHELB-VAWYXSNFSA-N
Synonym chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone
PubChem CID 637760
ChEBI CHEBI:48965
SMILES O=C(\C=C\C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 53.0°C to 58.0°C
Color Yellow
Boiling Point 345.0°C to 348.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CH=CHCOC6H5
Quantity 100 g
Beilstein 07, 478
Merck Index 15, 2036
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ether,chloroform,benzene and,carbon disulfide,slightly soluble in alcohol
IUPAC Name (2E)-1,3-diphenylprop-2-en-1-one
Formula Weight 208.26
Percent Purity 97%
Physical Form Granular Powder
Chemical Name or Material Chalcone, 97%
Show More Show Less
EINECSNumber 202-330-2
RTECSNumber FL6900000
TSCA TSCA

RUO – Research Use Only