Learn More
Excelta™ Chain Nose Pliers
Chain Nose Pliers with radiused edges to reduce scratching and marring of delicate leads. Made of AISI 440 Stainless Steel.
Supplier:  Excelta™ 2844
 View more versions of this product
Catalog No. 17467404
Only null left
Description
Description
  • Ergonomically designed, ESD-safe handles
Specifications
Chemical Identifiers
Chain Nose Pliers
Stainless Steel
Specifications
Chain Nose Pliers
5 in.
32.5 mm
ESD-safe Soft Foam Cushion
1.3 in.
Fine Precision
12.7 cm
Stainless Steel
Serrated/Non-serrated
Medium Chain Nose
Product Suggestions
Product Suggestions