missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Excelta™ Chain Nose Pliers
Chain Nose Pliers with radiused edges to reduce scratching and marring of delicate leads. Made of AISI 440 Stainless Steel.
Supplier: Excelta™ 2644S
Description
- Ergonomically designed, ESD-safe handles
Chemical Identifiers
| Chain Nose Pliers | |
| Stainless Steel |
Specifications
| Chain Nose Pliers | |
| 6 in. | |
| 22.5 mm | |
| Long 'S' Grip, ESD-safe Soft Foam Cushion | |
| 1.3 in. |
| Fine Precision | |
| 15.24 cm | |
| Stainless Steel | |
| Smooth | |
| Small Chain Nose |