missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Excelta™ Chain Nose Pliers
Chain Nose Pliers with radiused edges to reduce scratching and marring of delicate leads. Made of AISI 440 Stainless Steel.
Supplier: Excelta™ 11DI
Description
- Ergonomically designed, ESD-safe handles
Chemical Identifiers
| Chain Nose Pliers | |
| Stainless Steel |
Specifications
| Chain Nose Pliers | |
| 4.75 in. | |
| 30 mm | |
| ESD-safe Soft Foam Cushion | |
| 0.9 in. |
| High Precision | |
| 12 cm | |
| Stainless Steel | |
| Serrated | |
| Medium Chain Nose, Flame Hardened |