missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic Acid Sodium Salt, Monohydrate, Reagent HPLC, 99%, Spectrum™ Chemical

Catalog No. 18600071
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
5 kg
Packaging:
Amber Glass Bottle
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600071 5 kg Fiber Drum
18600068 5 g Amber Glass Bottle
18600069 25 g Amber Glass Bottle
18600070 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600071 Supplier Spectrum Chemical Mfg Cor Supplier No. O10015KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

O1001, 207596-29-0, C8H17NaO3S·H2O

Spectrum™ Chemical 1-Octanesulfonic Acid Sodium Salt, Monohydrate, Reagent HPLC is an ion pairing reagent which will enhance the retention of polar, ionizable compounds such as peptides and proteins. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 207596-29-0
Molecular Formula C8H19NaO4S
Molecular Weight (g/mol) 234.29
MDL Number MFCD00149551
InChI Key MBURIAHQXJQKRE-UHFFFAOYSA-M
IUPAC Name sodium hydrate octane-1-sulfonate
SMILES O.[Na+].CCCCCCCCS([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Fiber Drum
Quantity 5 kg
Percent Purity 99%
Grade Reagent