missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CGI-09 Recombinant Protein Antigen

Catalog No. NBP258675PE
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP258675PE 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP258675PE Supplier R&D Systems™ Supplier No. NBP258675PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CGI-09. Source: E.coli Amino Acid Sequence: LKDKGIKGEEIVQQLIENSTTFRDKTEFAQDKYIKKKKKKYEAIITVVKPSTRILSIMYYAREPGKINHMRYDTLAQMLTLGNIRAGN The CGI-09 Recombinant Protein Antigen is derived from E. coli. The CGI-09 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 51605
Product Line Novus
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name CGI-09 Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias CGI-09, GCD10, Gcd10p, KIAA1153, MGC5029, TRM6, tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6, tRNA methyltransferase 6 homolog (S. cerevisiae), tRNA(m1A58)-methyltransferase subunit TRM6, tRNA(m1A58)MTase subunit TRM6
Gene Symbol TRMT6
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 100 μL
Regulatory Status RUO
Source E.coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-51673. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only.