Learn More
ACROBiosystems Inc ACROBiosystems Anti-Cetuximab Antibodies (Non-Neutralizing)
Catalog No. 501956992
Encompass
Antibody;1MG;IgG1/kappa;Recognizes Cetuximab specifically, no cross reactivity with other humanized antibodies.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab')2.; ;Anti-Cetuximab Antibodies bridging ELISA for Anti-Drug Antibody (ADA) assay development.Immobilized cetuximab at 1 g/mL, add increasing concentrations of Anti-Cetuximab Antibodies (Cat. No. CEB-Y31, 10% human serum) and then add biotinylated cetuximab at 2 g/mL. Detection was performed using HRP-conjugated streptavidin with a sensitivity of 39 ng/mL. ;Demonstration of the specificity of Anti-Cetuximab Antibodies (Cat. No. CEB-Y31) to the cetuximab.;Anti-Cetuximab Antibodies (mouse IgG1, Cat. No. CEB-Y31) captured on CM5 chip via anti-mouse antibodies surface, can bind human cetuximab with an affinity constant of 0.421 nM.
Catalog No. 501956992 Supplier ACROBiosystems Inc Supplier No. CEBY311MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Antibody;1MG;IgG1/kappa;Recognizes Cetuximab specifically, no cross reactivity with other humanized antibodies.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab')2.; ;Anti-Cetuximab Antibodies bridging ELISA for Anti-Drug Antibody (ADA) assay development.Immobilized cetuximab at 1 g/mL, add increasing concentrations of Anti-Cetuximab Antibodies (Cat. No. CEB-Y31, 10% human serum) and then add biotinylated cetuximab at 2 g/mL. Detection was performed using HRP-conjugated streptavidin with a sensitivity of 39 ng/mL. ;Demonstration of the specificity of Anti-Cetuximab Antibodies (Cat. No. CEB-Y31) to the cetuximab.;Anti-Cetuximab Antibodies (mouse IgG1, Cat. No. CEB-Y31) captured on CM5 chip via anti-mouse antibodies surface, can bind human cetuximab with an affinity constant of 0.421 nM.