missing translation for 'onlineSavingsMsg'
Learn More

Cesium tungsten oxide, 99.9% (metals basis)

Catalog No. AA3920606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3920606 5 g
AA3920614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3920606 Supplier Thermo Scientific Chemicals Supplier No. 03920606
Only null left

CAS: 13587-19-4 | Cs2O4W

Cesium tungsten oxide is used for making transparent insulation coating that has the best infrared transparent barrier feature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13587-19-4
Molecular Formula Cs2O4W
MDL Number MFCD00049438
Synonym Cesium tungstate

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information It is soluble in water.
Formula Weight 513.66
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cesium tungsten oxide
EINECSNumber 237-019-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only