missing translation for 'onlineSavingsMsg'
Learn More

Cesium titanium oxide, 99.85% (metals basis)

CAS: 12158-57-5

Supplier:  Thermo Scientific Chemicals 03920714

 View more versions of this product

Catalog No. AA3920714

Only null left
Explore available promotions

Description

Description

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cesium titanium oxide
12158-57-5
25 g
Cs2TiO3
Cesium titanate
361.71
White
Powder
(metals basis)
MFCD00053969
Insoluble in water.
99.85%

Chemical Identifiers

12158-57-5
MFCD00053969
Cesium titanate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 235-283-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only