missing translation for 'onlineSavingsMsg'
Learn More

Cesium Pivalate 97.0+%, TCI America™

Catalog No. C32305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32305G 5 g
C32301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32305G Supplier TCI America Supplier No. C32305G
Only null left

Chemical Identifiers

CAS 20442-70-0
Molecular Formula C5H9CsO2
Molecular Weight (g/mol) 234.03
MDL Number MFCD10567040
InChI Key LGVUAXNPXVXCCW-UHFFFAOYSA-M
Synonym Pivalic Acid Cesium Salt, Cesium Trimethylacetate, Trimethylacetic Acid Cesium Salt, Cesium 2,2-Dimethylpropionate, 2,2-Dimethylpropionic Acid Cesium Salt
PubChem CID 583616
IUPAC Name caesium(1+) 2,2-dimethylpropanoate
SMILES [Cs+].CC(C)(C)C([O-])=O

Specifications

Melting Point 348°C
Color White
Quantity 5 g
Formula Weight 234.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cesium Pivalate