Learn More
Cesium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Description
Cesium perchlorate is used for long-duration optical transmission of information with infofuses. It is a modifier of polymer electrolyte/luminescent conjugated polymer mixed ionic-electronic conductors. It has been used as a reactant for investigating metal salt effects on morphology of silica nanomaterials prepared by polymerization on hydrogels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 250°C (decomposition) |
| Physical Form | Powder |
| Quantity | 25 g |
| UN Number | UN1481 |
| Solubility Information | Soluble in water. |
| Formula Weight | 232.35 |
| Grade | Reagent |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Cesium perchlorate |
Safety and Handling
| Hazard Category | H272-H315-H319-H335 |
| Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S. |
| EINECSNumber | 236-643-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only