missing translation for 'onlineSavingsMsg'
Learn More

Cesium Iodide, Approx. 99.9%, Spectrum™ Chemical

Catalog No. p-7165382
Change view
Click to view available options
Quantity:
25 g
100 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609638 25 g Amber Glass Bottle
18609637 100 g Amber Glass Bottle
18609639 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609638 Supplier Spectrum Chemical Mfg Cor Supplier No. C148325GM
Only null left

Chemical Identifiers

CAS 7789-17-5

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity ∼99.9%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.