missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium iodide, 99.999%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Boiling Point | 1280.0°C |
| Melting Point | 626.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 10 g |
| Assay Percent Range | 99.999% |
| Linear Formula | CsI |
| Merck Index | 15,2013 |
| Solubility Information | Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol, slightly soluble in methanol, practically insoluble in acetone |
| IUPAC Name | cesium;iodide |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Suspected of damaging fertility or the unborn child if inhaled. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the |
| EINECSNumber | 232-145-2 |
RUO – Research Use Only