missing translation for 'onlineSavingsMsg'
Learn More

Cesium iodide, 99.9%, for analysis

Catalog No. p-222838
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192050250 25 g Glass bottle
AC192051000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC192050250 Supplier Thermo Scientific Chemicals Supplier No. 192050250
Only null left

CAS: 7789-17-5 | CsI | 259.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-17-5
Molecular Formula CsI
Molecular Weight (g/mol) 259.81
MDL Number MFCD00010962
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
PubChem CID 24601
IUPAC Name cesium;iodide
SMILES [I-].[Cs+]

Specifications

Boiling Point 1280.0°C
Melting Point 626.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99.9%
Linear Formula CsI
Merck Index 15, 2013
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol, slightly soluble in methanol, practically insoluble in acetone
Formula Weight 259.8
Percent Purity 99.9%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Cesium iodide, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging fertility or the unborn child if inhaled.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the
EINECSNumber 232-145-2
RTECSNumber FL0350000
TSCA TSCA

RUO – Research Use Only