missing translation for 'onlineSavingsMsg'
Learn More

Cesium(I) Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. p-7125910
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32641G 1 g
C32645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32641G Supplier TCI America Supplier No. C32641G
Only null left

Chemical Identifiers

CAS 91742-16-4
Molecular Formula C2CsF6NO4S2
Molecular Weight (g/mol) 413.041
InChI Key NDUUPYVMAUBBMO-UHFFFAOYSA-N
Synonym Bis(trifluoromethanesulfonyl)imide Cesium(I) Salt, Cesium(I) Triflimide
PubChem CID 101771832
IUPAC Name cesium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cs+]

Specifications

Melting Point 122°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 413.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cesium(I) Bis(trifluoromethanesulfonyl)imide