missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium(I) Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 122°C |
| Color | White |
| Quantity | 5 g |
| UN Number | 1759 |
| Formula Weight | 413.04 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cesium(I) Bis(trifluoromethanesulfonyl)imide |
Safety and Handling
| TSCA | No |