missing translation for 'onlineSavingsMsg'
Learn More

Cesium hydroxide, 50% w/w aq. soln., 99.9% (metals basis)

Catalog No. p-7048279
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10020ND 25 g
AA10020NF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10020ND Supplier Thermo Scientific Chemicals Supplier No. 010020ND

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21351-79-1 | CsHO | 149.91 g/mol

Cesium hydroxide is used in electric storage batteries. Also used to make porous cesium titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. Cesium Hydroxide is used as electrolyte in alkaline batteries at subzero temperatures, color photography and as a catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21351-79-1
Molecular Formula CsHO
Molecular Weight (g/mol) 149.91
MDL Number MFCD00010964
InChI Key HUCVOHYBFXVBRW-UHFFFAOYSA-M
Synonym cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution
PubChem CID 62750
ChEBI CHEBI:33988
IUPAC Name cesium;hydroxide
SMILES [OH-].[Cs+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 25 g
UN Number UN2681
Solubility Information Highly soluble in water.
Formula Weight 149.91
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Sensitivity Air Sensitive
Density 1.725 g/mL
Assay (metals basis)
Chemical Name or Material Cesium hydroxide
Show More Show Less
Hazard Category H302-H314-H335-H361
Hazard Statement GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CAESIUM HYDROXIDE SOLUTION
EINECSNumber 244-344-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only