missing translation for 'onlineSavingsMsg'
Learn More

Cesium Fluoride, TCI America™

Catalog No. C220425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C220425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C220425G Supplier TCI America Supplier No. C220425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13400-13-0
Molecular Formula CsF
Molecular Weight (g/mol) 151.90
MDL Number MFCD00010960
InChI Key XJHCXCQVJFPJIK-UHFFFAOYSA-M
Synonym cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
PubChem CID 25953
IUPAC Name caesium(1+) fluoride
SMILES [F-].[Cs+]

Specifications

Melting Point 602°C
Color White
Quantity 25 g
UN Number 2923
Formula Weight 151.90
Physical Form Crystalline Powder
Chemical Name or Material Cesium Fluoride
EINECSNumber (1)-0662
RTECSNumber FK9650000
TSCA Yes