Learn More
Cesium fluoride, Puratronic™, 99.99% (metals basis)
Description
Cesium fluoride is used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1251°C |
| Melting Point | 682°C |
| Color | White |
| pH | 6.5 to 7.5 |
| Physical Form | Crystalline |
| Quantity | 1 g |
| UN Number | UN2923 |
| Solubility Information | Soluble in water. Insoluble in dioxane and pyridine. |
| Formula Weight | 151.9 |
| Percent Purity | 99.99% |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331-H315-H318-H361 |
| Hazard Statement | GHS H Statement H300-H310-H330-H314-H318 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P332+P313-P361-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 236-487-3 |
| RTECSNumber | FK9650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only