missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents

Catalog No. p-25038
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
BP210500 500 g Poly Bottle
BP2101 1 kg Poly Bottle
2 options

Catalog No. BP210500

Supplier: Fisher BioReagents BP210 500

Only null left
This Molecular Biology-grade cesium chloride is ideal for UV analysis of nucleic acids during density-gradient centrifugation and for the isolation of plasmid DNA, phage DNA, virus particles, and RNA.

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range ≥99.999 %
Merck Index 15, 2010
Formula Weight 168.36
Percent Purity ≥99.999%
Grade DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Refractive Index 1.367 to 1.371 (at 25°C)
Chemical Name or Material Cesium Chloride
Show More Show Less
Recommended Storage RT