missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
$2307.22 - $13248.67
Chemical Identifiers
| CAS | 7647-17-8 |
|---|---|
| Molecular Formula | ClCs |
| Molecular Weight (g/mol) | 168.36 |
| MDL Number | MFCD00010955 |
| InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
| Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
| PubChem CID | 24293 |
| ChEBI | CHEBI:63039 |
| IUPAC Name | cesium;chloride |
| SMILES | [Cl-].[Cs+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02002568
|
Avantor J.T.Baker
404204 |
100 g | Poly bottle |
Each for $2,307.22
|
|
||||
|
02002569
|
Avantor J.T.Baker
404202 |
1 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- For Density Gradient Centrifugation; For Laboratory,Research,or Manufacturing Use
Chemical Identifiers
| 7647-17-8 | |
| 168.36 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| 24293 | |
| cesium;chloride |
| ClCs | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| CHEBI:63039 | |
| [Cl-].[Cs+] |
Specifications
| 7647-17-8 | |
| ClCs | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| [Cl-].[Cs+] | |
| 168.36 | |
| CHEBI:63039 | |
| Ultrapure Bioreagent | |
| 1L = 3.99kg |
| 100 g | |
| MFCD00010955 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| cesium;chloride | |
| 24293 | |
| 168.36 | |
| Poly bottle | |
| Cesium Chloride |