missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium chloride, ultrapure, 99.9% (metals basis)
CAS: 7647-17-8 | ClCs | 168.36 g/mol
$310.70 - $310.70
Chemical Identifiers
| CAS | 7647-17-8 |
|---|---|
| Molecular Formula | ClCs |
| Molecular Weight (g/mol) | 168.36 |
| MDL Number | MFCD00010955 |
| InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
| Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
| PubChem CID | 24293 |
| ChEBI | CHEBI:63039 |
| IUPAC Name | cesium;chloride |
| SMILES | [Cl-].[Cs+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6595022
|
Thermo Scientific Chemicals
J6595022 |
100 g |
Each for $310.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7647-17-8 | |
| 168.36 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| 24293 | |
| cesium;chloride |
| ClCs | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| CHEBI:63039 | |
| [Cl-].[Cs+] |
Specifications
| 1303°C | |
| 7647-17-8 | |
| Crystalline | |
| ClCs | |
| 14,2011 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| cesium;chloride | |
| 24293 | |
| 168.36 | |
| Odorless | |
| Hygroscopic | |
| Cesium chloride | |
| Ultrapure |
| 646°C | |
| White | |
| 100 g | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| [Cl-].[Cs+] | |
| 168.36 | |
| CHEBI:63039 | |
| 99.9% | |
| 1.6418 | |
| 3.99 | |
| (metals basis) |
Safety and Handling
P201-P202-P260-P281-P308+P313-P501c
H361-H373
EINECSNumber : 231-600-2
RTECSNumber : FK9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only