missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride 99.0+%, TCI America™
$83.89 - $216.24
Chemical Identifiers
| CAS | 7647-17-8 |
|---|---|
| Molecular Formula | ClCs |
| Molecular Weight (g/mol) | 168.36 |
| MDL Number | MFCD00010955 |
| InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
| Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
| PubChem CID | 24293 |
| ChEBI | CHEBI:63039 |
| IUPAC Name | caesium(1+) chloride |
| SMILES | [Cl-].[Cs+] |
Chemical Identifiers
| 7647-17-8 | |
| 168.36 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| 24293 | |
| caesium(1+) chloride |
| ClCs | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| CHEBI:63039 | |
| [Cl-].[Cs+] |
Specifications
| White | |
| 7647-17-8 | |
| ClCs | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| caesium(1+) chloride | |
| 24293 | |
| 168.36 | |
| Cesium Chloride |
| Crystal-Powder at 20°C | |
| 645°C | |
| MFCD00010955 | |
| 25 g | |
| [Cl-].[Cs+] | |
| 168.36 | |
| CHEBI:63039 | |
| ≥99.0% (T) |
Safety and Handling
EINECSNumber : (1)-0631
RTECSNumber : FK9625000
TSCA : Yes