missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Physical Form | Crystal-Powder at 20°C |
| Melting Point | 645°C |
| Quantity | 100 g |
| Formula Weight | 168.36 |
| Percent Purity | ≥99.0% (T) |
| Chemical Name or Material | Cesium Chloride |
Safety and Handling
| EINECSNumber | (1)-0631 |
| RTECSNumber | FK9625000 |
| TSCA | Yes |