missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Cesium Chloride, Molecular Biology Grade, Calbiochem™,
Used for density gradient centrifugation. Commonly used for fractionation of nucleic acids, ribosomal subunits, proteins, glycoproteins, and viruses
$377.00 - $1210.00
Chemical Identifiers
| CAS | 7647-17-8 |
|---|---|
| Molecular Formula | ClCs |
| Molecular Weight (g/mol) | 168.36 |
| MDL Number | MFCD00010955 |
| InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
| Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
| PubChem CID | 24293 |
| ChEBI | CHEBI:63039 |
| IUPAC Name | caesium(1+) chloride |
| SMILES | [Cl-].[Cs+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
219650100GM
|
MilliporeSigma™
219650100GM |
100 g |
Each for $377.00
|
|
|||||
|
219650500GM
|
MilliporeSigma™
219650500GM |
500 g |
Each for $1,210.00
|
|
|||||
Description
HygroscopicChemical Identifiers
| 7647-17-8 | |
| 168.36 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| 24293 | |
| caesium(1+) chloride |
| ClCs | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| CHEBI:63039 | |
| [Cl-].[Cs+] |
Specifications
| 7647-17-8 | |
| ≥99% | |
| MFCD00010955 | |
| Easily soluble in cold water | |
| [Cl-].[Cs+] | |
| 168.36 | |
| CHEBI:63039 | |
| Cesium Chloride |
| 100 g | |
| ClCs | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| caesium(1+) chloride | |
| 24293 | |
| 168.4 | |
| by titration (argentometry) |
Safety and Handling
EINECSNumber : 231-600-2
RTECSNumber : FK9625000
Recommended Storage : Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).