missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Cesium Chloride, Molecular Biology Grade, Calbiochem™,

Catalog No. 219650100GM
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
219650100GM 100 g
219650500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 219650100GM Supplier MilliporeSigma™ Supplier No. 219650100GM

Used for density gradient centrifugation. Commonly used for fractionation of nucleic acids, ribosomal subunits, proteins, glycoproteins, and viruses

Hygroscopic

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Quantity 100 g
Assay Percent Range ≥99%
Solubility Information Easily soluble in cold water
Formula Weight 168.4
Chemical Name or Material Cesium Chloride
Grade by titration (argentometry)
EINECSNumber 231-600-2
RTECSNumber FK9625000
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).