missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride (Crystalline Powder), Fisher BioReagents

Catalog No. p-25029
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
BP1595500 500 g Poly Bottle
BP15951 1 kg Poly Bottle
2 options

Catalog No. BP1595500

Supplier: Fisher BioReagents BP1595500

Only null left

For isolation of phage DNA, plasmid DNA, or total RNA

This economical grade of Cesium Chloride can be used where low levels of metal ion impurities do not interfere. It is recommended for isolating phage DNA, plasmid DNA, or total RNA.

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥99.5 %
Merck Index 15, 2010
Formula Weight 168.36
Percent Purity ≥99.5%
Packaging Poly Bottle
Refractive Index 1.364 to 1.374 (at 25°C)
Chemical Name or Material Cesium Chloride
Grade DNase-, RNase- and Protease-Free
Show More Show Less
Recommended Storage RT