missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride (Crystalline Powder), Fisher BioReagents

Catalog No. BP15951
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP15951 1 kg Poly Bottle
BP1595500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP15951 Supplier Fisher BioReagents Supplier No. BP15951
Only null left

For isolation of phage DNA, plasmid DNA, or total RNA

This economical grade of Cesium Chloride can be used where low levels of metal ion impurities do not interfere. It is recommended for isolating phage DNA, plasmid DNA, or total RNA.

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Powder/Solid
Quantity 1 kg
Assay Percent Range ≥99.5 %
Merck Index 15, 2010
Formula Weight 168.36
Percent Purity ≥99.5%
Packaging Poly Bottle
Refractive Index 1.364 to 1.374 (at 25°C)
Chemical Name or Material Cesium Chloride
Grade DNase-, RNase- and Protease-Free
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT