missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99+%, pure

Catalog No. p-3756625
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC422850250 25 g Glass bottle
AC422851000 100 g Glass bottle
2 options

Catalog No. AC422850250

Supplier: Thermo Scientific Chemicals 422850250

Only null left

CAS: 7647-17-8 | ClCs | 168.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290.0°C
Melting Point 646.0°C
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula CsCl
Merck Index 15,201
Solubility Information Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
Formula Weight 168.36
Percent Purity ≥99%
Grade Pure
Packaging Glass bottle
Density 3.983g/mL
Chemical Name or Material Cesium chloride, Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 231-600-2

RUO – Research Use Only