missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99+%, for analysis

Catalog No. p-46317
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC189502500 250 g Plastic bottle
AC189500010 1 kg Plastic bottle
2 options

Catalog No. AC189502500

Supplier: Thermo Scientific Chemicals 189502500

Only null left

CAS: 7647-17-8 | ClCs | 168.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290.0°C
Melting Point 646.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula CsCl
Merck Index 15, 2010
Solubility Information Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
IUPAC Name caesium(1+) chloride
Formula Weight 168.36
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 3.983
Chemical Name or Material Cesium chloride, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 231-600-2
RTECSNumber FK9625000
TSCA TSCA

RUO – Research Use Only