missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, 99.99%, (Optical Grade I), MP Biomedicals

Catalog No. ICN16004125
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16004125 25 g
ICN16004180 100 g
ICN16004190 500 g
ICN16004191 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN16004125 Supplier MP Biomedicals Inc Supplier No. 0216004125
Only null left

Solutions are clear and colorless suitable for density gradient work, nucleic acid and protein purification

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Quantity 25 g
Assay Percent Range 99.99%
Formula Weight 168.4
Grade Optical I
Chemical Name or Material Cesium Chloride
Recommended Storage Room Temperature