missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Bromide 99.0+%, TCI America™
Description
- Inorganic cesium salt used primarily as a reagent in research
Specifications
Specifications
| Melting Point | 636°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.00% |
| Merck Index | 2009 |
| Formula Weight | 212.81 |
| Percent Purity | ≥99.0% (T) |
| Sensitivity | Hygroscopic |
| Appearance | White Powder to Crystal |
| Show More |
Safety and Handling
| EINECSNumber | (1)-1245 |
| RTECSNumber | FK9275000 |
| TSCA | Yes |