missing translation for 'onlineSavingsMsg'
Learn More

Cesium Bromide 99.0+%, TCI America™

Catalog No. C2202100G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2202100G 100 g
C220225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2202100G Supplier TCI America Supplier No. C2202100G
Only null left

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
PubChem CID 24592
IUPAC Name caesium(1+) bromide
SMILES [Br-].[Cs+]

Specifications

Melting Point 636°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 212.81
Percent Purity ≥99.0% (T)
Chemical Name or Material Cesium Bromide
EINECSNumber (1)-1245
RTECSNumber FK9275000
TSCA Yes