missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Acetate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 193°C |
| Color | White |
| Quantity | 100 g |
| Formula Weight | 191.95 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Cesium Acetate |
Safety and Handling
| TSCA | Yes |