missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium acetate, 99%, pure, Thermo Scientific™
$121.01 - $457.27
Chemical Identifiers
| CAS | 3396-11-0 |
|---|---|
| Molecular Formula | C2H3CsO2 |
| Molecular Weight (g/mol) | 191.96 |
| MDL Number | MFCD00013056 |
| InChI Key | ZOAIGCHJWKDIPJ-UHFFFAOYSA-M |
| Synonym | cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion |
| PubChem CID | 5152919 |
| IUPAC Name | cesium;acetate |
| SMILES | CC(=O)[O-].[Cs+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC191040250
|
Thermo Scientific Chemicals
191040250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC191041000
|
Thermo Scientific Chemicals
191041000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 3396-11-0 | |
| 191.96 | |
| ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
| 5152919 | |
| CC(=O)[O-].[Cs+] |
| C2H3CsO2 | |
| MFCD00013056 | |
| cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
| cesium;acetate |
Specifications
| 3396-11-0 | |
| Glass bottle | |
| CH3CO2Cs | |
| 25g | |
| Solubility in water: 91.5% (21°C). | |
| CC(=O)[O-].[Cs+] | |
| 191.96 | |
| 191.96 | |
| Pure |
| 99% | |
| C2H3CsO2 | |
| MFCD00013056 | |
| cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
| ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
| cesium;acetate | |
| 5152919 | |
| 99% | |
| Cesium acetate, Pure |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.
GHS Signal Word: Warning
EINECSNumber : 222-248-
RUO â Research Use Only