missing translation for 'onlineSavingsMsg'
Learn More

Cesium acetate, 99.998% (metals basis)

Catalog No. p-7059336
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4443414 25 g
AA4443422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4443414 Supplier Thermo Scientific Chemicals Supplier No. 04443414
Only null left

CAS: 3396-11-0 | C2H3CsO2 | 191.949 g/mol

Cesium acetate is used in organic synthesis especially in the Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.949
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194°C
Physical Form Lump/Powder
Quantity 25 g
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 191.95
Percent Purity 99.998%
Odor Weak
Assay (metals basis)
Chemical Name or Material Cesium acetate
Hazard Category H361
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 222-248-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only