Learn More
StatLab Cervical Broom Sterile
A device designed to gather both endo and ectocervical cells including those in the transformation zone.
Supplier:  StatLab CB400100
Catalog No. STLCB400100
Only null left
Explore available promotions
Description
Description
  • Unique, softer LDP bristles with slip agent gather full spectrum of cells in a one-step procedure
  • Sterile, individually packaged
  • 100 per package
Specifications
Specifications
Cervex-Brush Combination
Polyethylene
Sterile
Yes
Product Suggestions
Product Suggestions