missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Certified Dumbbells

Catalog No. p-8401019
Encompass
Change view
Click to view available options
Size:
Medium
Length (English):
4.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English)
14727008 Medium 4.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14727008 Supplier Bio-Serv™ Supplier No. K3224
Only null left

Large end knobs minimize loss through cage bars and floor slats

  • Made of solid polypropylene
  • Large ends knobs minimize loss through cage bars and floor slats
  • Extremely durable
  • Autoclavable
  • Cagewasher safe

Specifications

Autoclavable Autoclavable
Color Assorted
Material Polypropylene
For Use With (Application) Primiate Enrichment
Size Medium
Length (English) 4.75 in.