missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ Certified Dumbbells
Description
- Made of solid polypropylene
- Large ends knobs minimize loss through cage bars and floor slats
- Extremely durable
- Autoclavable
- Cagewasher safe
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Assorted |
| Material | Polypropylene |
| For Use With (Application) | Primiate Enrichment |
| Size | Medium |
| Length (English) | 4.75 in. |