Learn More
GE Analytical Instruments GE Certified Blue TOC Vials, 40-mL, 72/cs
Catalog No. NC1053336
Encompass
Pre-cleaned and certified (<10 ppb TOC) 40-mL glass vials with caps and Teflon-lined septa. Paper free packaging, produced in an ISO 8 Cleanroom. 72/cs
Catalog No. NC1053336 Supplier GE Analytical Instruments Supplier No. HMI 9061001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Pre-cleaned and certified (<10 ppb TOC) 40-mL glass vials with caps and Teflon-lined septa. Paper free packaging, produced in an ISO 8 Cleanroom. 72/cs