missing translation for 'onlineSavingsMsg'
Learn More

Cerotic Acid 95.0+%, TCI America™

$290.79 - $2093.33

Chemical Identifiers

CAS 506-46-7
Molecular Formula C26H52O2
Molecular Weight (g/mol) 396.7
MDL Number MFCD00002811
InChI Key XMHIUKTWLZUKEX-UHFFFAOYSA-N
Synonym cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
PubChem CID 10469
ChEBI CHEBI:31009
IUPAC Name hexacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C08291G
View Documents
TCI America
C08291G
1 g
Each for $290.79
Only null left
 
C082910G
View Documents
TCI America
C082910G
10 g
Each for $2,093.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

506-46-7
396.7
XMHIUKTWLZUKEX-UHFFFAOYSA-N
10469
hexacosanoic acid
C26H52O2
MFCD00002811
cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
CHEBI:31009
CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

506-46-7
White-Yellow
MFCD00002811
cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
396.7
CHEBI:31009
≥95.0% (GC,T)
Cerotic Acid
88°C
C26H52O2
1 g
XMHIUKTWLZUKEX-UHFFFAOYSA-N
hexacosanoic acid
10469
396.70
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents