missing translation for 'onlineSavingsMsg'
Learn More

Cerium zirconium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7050655
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3921622 100 g
AA3921636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3921622 Supplier Thermo Scientific Chemicals Supplier No. 03921622
Only null left

Used as wear resistant grinding and dispersion media. Used in fuel cell materials (SOFC), body armors. Also finds applications in catalytic converter, honey comb, precision ceramic parts, abrasive wheels, oxygen sensors, cutting tools. Used as ceramic steel. Other uses include screen printing, dip and spin coating and APS coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53169-24-7
MDL Number MFCD00168089
Synonym Cerium zirconate

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 295.32
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Cerium zirconium oxide
EINECSNumber 258-407-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only